missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CGS 21680 hydrochloride
GREENER_CHOICE

Product Code. 30001186
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001186 10 mg 10mg
18094162 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001186 Supplier Tocris Bioscience™ Supplier No. 1063/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A2A agonist

CGS 21680 hydrochloride is a potent A2A adenosine receptor agonist (Ki = 27 nM, EC50 = 1.48-180 nM). CGS 21680 reduces venous resistance, increases heart rate and cardiac output in vivo. In an animal model of Huntington's disease, CGS21680 slows motor deterioration, prevents reduction in brain weight and reduces the size of mutant huntingtin aggregates and intranuclear inclusions. CGS21680 increases proliferation of neural progenitor cells, promotes survival of motoneurons and activates TrkB receptors. Also has anti-inflammatory effects in animal models of lung injury and reduces neurotoxicity induced by Kainate (Cat. No. 7065).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124431-80-7
Molecular Formula C23H30ClN7O6

Specifications

Quantity 10 mg
Formula Weight 535.99
Purity >98%
Molecular Weight (g/mol) 535.986
Chemical Name or Material CGS 21680 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.