missing translation for 'onlineSavingsMsg'
Learn More

CGS 21680A, TRC

Product Code. 30363162
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363162 10 mg 10mg
30362249 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30363162 Supplier Toronto Research Chemicals Supplier No. TRCC29179010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124431-80-7
Molecular Formula C23 H29 N7 O6 . Cl H
Molecular Weight (g/mol) 535.98
Synonym CGS 21680 HCl
IUPAC Name 3-[4-[2-[[6-amino-9-[(2R,3R,4S,5S)-5-(ethylcarbamoyl)-3,4-dihydroxyoxolan-2-yl]purin-2-yl]amino]ethyl]phenyl]propanoic acid;hydrochloride
SMILES Cl.CCNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)nc(NCCc4ccc(CCC(=O)O)cc4)nc23

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H29N7O6.ClH/c1-2-25-21(35)18-16(33)17(34)22(36-18)30-11-27-15-19(24)28-23(29-20(15)30)26-10-9-13-5-3-12(4-6-13)7-8-14(31)32;/h3-6,11,16-18,22,33-34H,2,7-10H2,1H3,(H,25,35)(H,31,32)(H3,24,26,28,29);1H/t16-,17+,18-,22+;/m0./s1
Formula Weight 535.1946
Chemical Name or Material CGS 21680A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.