missing translation for 'onlineSavingsMsg'
Learn More

CGS 21680 Hydrochloride, MedChemExpress

Product Code. 16455548
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455548 10 mg 10mg
16446106 100 mg 100mg
16441717 50 mg 50mg
16428257 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16455548 Supplier Medchem Express Supplier No. HY13201A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CGS 21680 Hydrochloride is a selective adenosine A2A receptor agonist with a Ki of 27 nM.

CGS 21680 Hydrochloride is a selective adenosine A2A receptor agonist with a Ki of 27 nM.

Specifications

Chemical Name or Material CGS 21680 Hydrochloride
CAS 124431-80-7
Color White
Purity Grade Notes Research
Molecular Formula C23H30ClN7O6
Quantity 10 mg
Solubility Information DMSO : ≥ 20 mg/mL (37.31 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)CCC1=CC=C(CCNC2=NC3=C(N=CN3[C@H]4[C@H](O)[C@H](O)[C@@H](C(NCC)=O)O4)C(N)=N2)C=C1.Cl
Molecular Weight (g/mol) 535.98
Formula Weight 535.98
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.