CAS RN 1222998-36-8
CAS RN 1222998-36-8
IUPAC Name:
1-[4-(4-propanoylpiperazin-1-yl)-3-(trifluoromethyl)phenyl]-9-(quinolin-3-yl)-1H,2H-benzo[h]1,6-naphthyridin-2-one
Synonyms:
1-[4-(4-propanoylpiperazin-1-yl)-3-(trifluoromethyl)phenyl]-9-(quinolin-3-yl)benzo[h]1,6-naphthyridin-2-one
Molecular Weight (g/mol):
607.64
Molecular Formula:
C35H28F3N5O2
InChi Key:
AKCRNFFTGXBONI-UHFFFAOYSA-N
SMILES:
CCC(=O)N1CCN(CC1)C1=C(C=C(C=C1)N1C(=O)C=CC2=C1C1=CC(=CC=C1N=C2)C1=CN=C2C=CC=CC2=C1)C(F)(F)F
1
–
3
of
3
results
1
Tocris Bioscience™ Torin 1
CAS: 1222998-36-8 Molecular Formula: C35H28F3N5O2
2
3
Torin 1, MedChemExpress
MedChemExpress Torin 1 is a potent inhibitor of mTOR with an IC50 of 3 nM. Torin 1 inhibits both mTORC1/2 complexes with IC50 values between 2 and 10 nM. Torin 1 is an effective inducer of autophagy.