missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Torin 1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Torin 1 is a potent and selective ATP-competitive mTOR inhibitor (IC50 = 2 - 10 nM for mTORC1 and mTORC2). Torin 1 displays 200-fold selectivity for mTOR over DNA-PK, ATM and hVps34. In HeLa cells, Torin 1 induces autophagy. Torin 1 impairs cell growth and proliferation by suppression of the rapamycin-resistant functions of mTORC1. In human monocytes and myeloid dendritic cells, Torin 1 prevents decreases the anti-inflammatory potency of glucocorticoids. In the human endocrine cell line BON, Torin 1 increases neurotensin secretion and gene expression through MEK/ERK/c-Jun pathway activation.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 607.62 |
| Purity | >98% |
| Molecular Weight (g/mol) | 607.64 |
| Chemical Name or Material | Torin 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction