CAS RN 119793-66-7
CAS RN 119793-66-7
IUPAC Name:
[(2R)-3-carboxy-2-(propanoyloxy)propyl]trimethylazanium chloride
Synonyms:
propionyl-L-carnitine chloride
Molecular Weight (g/mol):
253.72
Molecular Formula:
C10H20ClNO4
InChi Key:
KTFMPDDJYRFWQE-DDWIOCJRSA-N
SMILES:
[Cl-].CCC(=O)O[C@H](CC(O)=O)C[N+](C)(C)C
1
–
3
of
3
results
1
(R)-Propionyl Carnitine Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-Propionyl Carnitine Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Levocarnitine propionate hydrochloride, MedChemExpress
MedChemExpress Levocarnitine propionate hydrochloride (L-Propionylcarnitine chloride; ST-261) is used to treat the deterioration of renal function, congestive heart failure, intermittent claudication, and other diseases.