missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-Propionyl Carnitine Chloride, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | (R)-Propionyl Carnitine chloride |
| CAS | 119793-66-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H20 N O4 . Cl |
| Quantity | 25 mg |
| Synonym | 1-Propanaminium','3-carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-','chloride (1:1)','(2R)- (ACI), 1-Propanaminium','3-carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-','chloride','(2R)- (9CI), 1-Propanaminium','3-carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-','chloride','(R)- (ZCI), Dromos, Levocarnitine propionate hydrochloride, ST 261, (R)-Propionyl carnitine chloride, Propionyl-L-carnitine hydrochloride, (2R)-3-Carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-1-propanaminium chloride |
| InChI Formula | InChI=1S/C10H19NO4.ClH/c1-5-10(14)15-8(6-9(12)13)7-11(2,3)4;/h8H,5-7H2,1-4H3;1H/t8-;/m1./s1 |
| SMILES | [Cl-].CCC(=O)O[C@H](CC(=O)O)C[N+](C)(C)C |
| IUPAC Name | [(2R)-3-carboxy-2-propanoyloxypropyl]-trimethylazanium;chloride |
| Molecular Weight (g/mol) | 253.72 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction