missing translation for 'onlineSavingsMsg'
Learn More

Levocarnitine propionate hydrochloride, MedChemExpress

Product Code. 16474507
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16474507

Brand: Medchem Express HYB0932

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Levocarnitine propionate hydrochloride (L-Propionylcarnitine chloride; ST-261) is used to treat the deterioration of renal function, congestive heart failure, intermittent claudication, and other diseases.

Levocarnitine propionate hydrochloride (L-Propionylcarnitine chloride

Specifications

Chemical Name or Material Levocarnitine propionate hydrochloride
CAS 119793-66-7
Color White
Purity Grade Notes Research
Molecular Formula C10H20ClNO4
Quantity 50 mg
Synonym L-Propionylcarnitine chloride ST-261
Solubility Information DMSO : 250 mg/mL (985.34 mM; Need ultrasonic) ∣H2O : 60 mg/mL (236.48 mM; Need ultrasonic)
SMILES CCC(O[C@H](CC(O)=O)C[N+](C)(C)C)=O.[Cl-]
Molecular Weight (g/mol) 253.72
Formula Weight 253.72
Percent Purity 95.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.