accessibility menu, dialog, popup

UserName

CAS RN 107516-75-6

CH3OOHNOOH3C

CAS RN 107516-75-6

IUPAC Name: 2,6-diethyl 1H-indole-2,6-dicarboxylate
Synonyms: 2,6-diethyl 1H-indole-2,6-dicarboxylate
Molecular Weight (g/mol): 261.28
Molecular Formula: C14H15NO4
InChi Key: ISQYBMUDGXFHAF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC=C(C=C2N1)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results