missing translation for 'onlineSavingsMsg'
Learn More

Diethyl indole-2,6-dicarboxylate, 96%, Thermo Scientific™

Product Code. 15410868
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410868 250 mg 250mg
15400868 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15410868 Supplier Thermo Scientific Alfa Aesar Supplier No. H54619.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107516-75-6
Molecular Formula C14H15NO4
Molecular Weight (g/mol) 261.277
MDL Number MFCD03411566
InChI Key ISQYBMUDGXFHAF-UHFFFAOYSA-N
Synonym 6-ethoxycarbonylindole-2-carboxylic acid ethyl ester, diethyl indole-2,6-dicarboxylate, 1h-indole-2,6-dicarboxylic acid diethyl ester, 2,6-diethyl 1h-indole-2,6-dicarboxylate, pubchem15804, dethyl indole-2,6-dicarboxylate, indole-2,6-dicarboxylic acid diethyl ester, 6-ethoxycarbonylindole-2-carboxylic acid ethyl est
PubChem CID 44754779
IUPAC Name diethyl 1H-indole-2,6-dicarboxylate
SMILES CCOC(=O)C1=CC2=C(C=C1)C=C(N2)C(=O)OCC

Specifications

Melting Point 119°C to 121°C
Quantity 250 mg
Formula Weight 261.27
Percent Purity 96%
Chemical Name or Material Diethyl indole-2,6-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.