missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 200 μL

Product Code. 12756769 Shop All Axygen Scientific Products
Change view
Click to view available options
Tip Style:
Maxymum Recovery, Round
Standard, Round
Unit Size:
Pack of 4800
Pack of 960
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tip Style Unit Size
12756769 Standard, Round Pack of 4800
11384335 Standard, Round Pack of 960
11374335 Maxymum Recovery, Round Pack of 960
12746769 Maxymum Recovery, Round Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12756769 Supplier Axygen™ Supplier No. ZTF200RS
  • ONLINE EXCLUSIVE PRICE
    £733.00
    £716.46 / Pack of 4800
    Save £16.54
    2% Off
Estimated Shipment: 10-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Clear, autoclavable tips; color-coded racks.

  • Clear Tips
  • Format: 96 well
  • Propylene construction
  • Certified RNase/DNase free
  • Nonpyrogenic
  • Aerosol resistant filters
  • Sterile
  • Compatibility: Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head, Zymark™/Caliper RapidPlate™/SciClone™ with HV96 head/CAS-4200-1ch.-20-150μL/sec. head

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Blue
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
For Use With (Application) Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss
Format Rack
Tip Style Standard, Round
Packaging Non-SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Product Line Caliper Zymark
Material Polyethylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.