missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZXDC Monoclonal Antibody (K16280_15D3), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | ZXDC |
|---|---|
| Clone | K16280_15D3 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30205833
|
Invitrogen™
MA556058 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30195017
|
Invitrogen™
MA555678 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: SDDPLALTSN SNLAAHITTP TSSSTPRENA SVPELLAPIK VEPDSPSRPG AVGQQEGSHG LPQSTLPSPA EQHGAQDTEL SAGTGNFYLE SGGSARTDYR AIQLAKEKKQ RGAGSNAGAS QSTQRKIKEG
ZXDC Monoclonal antibody specifically detects ZXDC in Human, Mouse samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| ZXDC | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| A930012H20; B930086F11Rik; BC003332; LOW QUALITY PROTEIN: zinc finger protein ZXDC; RGD1561296; SERH2790; zinc finger protein ZXDC; ZXD family zinc finger C; ZXD family zinc finger C-like; ZXDC; ZXDL; ZXD-like zinc finger protein | |
| Recombinant human ZXDC (immunogen range is 621-750). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ZXDC |
| K16280_15D3 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q2QGD7, Q8C8V1 | |
| 79364, 80292 | |
| IgG2a | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title