missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZXDC Monoclonal Antibody (K16280_15D3), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA555678

 View more versions of this product

Product Code. 30195017

  • £416.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: SDDPLALTSN SNLAAHITTP TSSSTPRENA SVPELLAPIK VEPDSPSRPG AVGQQEGSHG LPQSTLPSPA EQHGAQDTEL SAGTGNFYLE SGGSARTDYR AIQLAKEKKQ RGAGSNAGAS QSTQRKIKEG

ZXDC belongs to the ZXD family. It contains 10 C2H2-type zinc fingers. ZXDC cooperates with CIITA to promote transcription of MHC class I and MHC class II genes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZXDC
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q2QGD7, Q8C8V1
ZXDC
Recombinant human ZXDC (immunogen range is 621-750).
50 μL
Primary
Human, Mouse
Antibody
IgG2a
Immunoprecipitation, Western Blot
K16280_15D3
Unconjugated
ZXDC
A930012H20; B930086F11Rik; BC003332; LOW QUALITY PROTEIN: zinc finger protein ZXDC; RGD1561296; SERH2790; zinc finger protein ZXDC; ZXD family zinc finger C; ZXD family zinc finger C-like; ZXDC; ZXDL; ZXD-like zinc finger protein
Mouse
Protein A
RUO
79364, 80292
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.