missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zuberitamab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen Zuberitamab Chimeric
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30282567
GREENER_CHOICE
View Documents
Invitrogen™
MA559189
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282563
View Documents
Invitrogen™
MA559190
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Zuberitamab, also known by its code name HS-006 and brand name Anruixi, is a biosimilar of rituximab that targets the CD20 antigen on B-cells. The CD20 gene, also known as MS4A1, encodes a B cell-specific membrane-bound protein that plays a crucial role in B cell development and function. Located on chromosome 11q12 in humans, the MS4A1 gene spans 16 kb and includes eight exons. CD20 is a part of the membrane-spanning 4-domain family, subfamily A (MS4A), which encompasses 18 tetraspan proteins. The prominent CD20 mRNA variant is 2.8 kb long, although other shorter variants exist. This phosphoprotein, approximately 33-37 kDa, is expressed on the surface of almost all normal and malignant B cells, making it an ideal target for therapeutic antibodies like rituximab. CD20 is known to interact with several proteins such as CD40, MHCII, and CD81, playing a role in B cell receptor signaling and other immune processes. Despite its widespread clinical use in B cell depletion therapies, the precise biological function of CD20 remains partly unresolved.
Specifications

Specifications

Zuberitamab Chimeric
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.