missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Zuberitamab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559190
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Zuberitamab, also known by its code name HS-006 and brand name Anruixi, is a biosimilar of rituximab that targets the CD20 antigen on B-cells. The CD20 gene, also known as MS4A1, encodes a B cell-specific membrane-bound protein that plays a crucial role in B cell development and function. Located on chromosome 11q12 in humans, the MS4A1 gene spans 16 kb and includes eight exons. CD20 is a part of the membrane-spanning 4-domain family, subfamily A (MS4A), which encompasses 18 tetraspan proteins. The prominent CD20 mRNA variant is 2.8 kb long, although other shorter variants exist. This phosphoprotein, approximately 33-37 kDa, is expressed on the surface of almost all normal and malignant B cells, making it an ideal target for therapeutic antibodies like rituximab. CD20 is known to interact with several proteins such as CD40, MHCII, and CD81, playing a role in B cell receptor signaling and other immune processes. Despite its widespread clinical use in B cell depletion therapies, the precise biological function of CD20 remains partly unresolved.
Specifications
| Zuberitamab Chimeric | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction