missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59436
This item is not returnable.
View return policy
Description
ZP2 Polyclonal specifically detects ZP2 in Human samples. It is validated for Western Blot.
Specifications
| ZP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Zona pellucida glycoprotein 2, zona pellucida glycoprotein 2 (sperm receptor), zona pellucida glycoprotein ZP2, Zona pellucida protein A, zona pellucida sperm-binding protein 2, Zp-2 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 7783 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q05996 | |
| ZP2 | |
| Synthetic peptides corresponding to ZP2(zona pellucida glycoprotein 2 (sperm receptor)) The peptide sequence was selected from the C terminal of ZP2. Peptide sequence PDSFPQWNVVVDGCAYDLDNYQTTFHPVGSSVTHPDHYQRFDMKAFAFVS. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Rabbit: 92%; Canine: 85%; Bovine: 78%; Equine: 78%; Pig: 78%. | |
| Human, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction