missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZO-1 Monoclonal Antibody (ZO1-1A12), Alexa Fluor™ 647
GREENER_CHOICE

Product Code. 15648028
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15648028 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15648028 Supplier Invitrogen™ Supplier No. MA339100A647

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZO-1 Monoclonal Antibody for ICC/IF

ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZO-1
Applications Immunocytochemistry
Classification Monoclonal
Clone ZO1-1A12
Concentration 1 mg/mL
Conjugate Alexa Fluor 647
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene TJP1
Gene Accession No. O97758, Q07157
Gene Alias DKFZp686M05161; I79_005485; MGC133289; OTTHUMP00000174520; Tight junction protein 1; tight junction protein 1 (zona occludens 1); tight junction protein ZO-1; Tjp1; zo1; ZO-1; ZO-1 MDCK; zona occludens 1; zona occludens protein 1; zonula occludens 1; zonula occludens 1 protein; zonula occludens protein 1
Gene Symbols TJP1
Host Species Mouse
Immunogen Human recombinant ZO-1 fusion protein encompassing amino acids 334-634.
Purification Method Affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403752, 7082
Target Species Canine, Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.