missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF804A Antibody (ZNF804A/8945), Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £351.00
Specifications
| Antigen | ZNF804A |
|---|---|
| Clone | ZNF804A/8945 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601795
|
Novus Biologicals
NBP3-23688-100ug |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699653
|
Novus Biologicals
NBP3-23688-20ug |
20 μg |
£178.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF804A Monoclonal antibody specifically detects ZNF804A in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ZNF804A | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| C2orf10, ZNF804A zinc finger protein 804A | |
| Recombinant full-length human ZNF804A protein | |
| Primary | |
| Store at 4°C. |
| ZNF804A/8945 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 91752 | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title