missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF804A Antibody (ZNF804A/8945), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23688-100ug
This item is not returnable.
View return policy
Description
ZNF804A Monoclonal antibody specifically detects ZNF804A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ZNF804A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| C2orf10, ZNF804A zinc finger protein 804A | |
| Recombinant full-length human ZNF804A protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| ZNF804A/8945 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 91752 | |
| Store at 4°C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction