missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF579 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | ZNF579 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF579 Polyclonal specifically detects ZNF579 in Human samples. It is validated for Western Blot.Specifications
| ZNF579 | |
| Polyclonal | |
| Rabbit | |
| Q8NAF0 | |
| 163033 | |
| Synthetic peptides corresponding to ZNF579 (zinc finger protein 579) The peptide sequence was selected from the C terminal of ZNF579. Peptide sequence LASYLRQHRRVHGPLSLLAPLPAAGKKDDKASGARNSAKGPEGGEGAECG. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| FLJ35453, zinc finger protein 579 | |
| ZNF579 | |
| IgG | |
| 60 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title