missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF521 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41174
This item is not returnable.
View return policy
Description
ZNF521 Polyclonal specifically detects ZNF521 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ZNF521 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ZNF521 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human ZNF521. The immunogen is located within the first 50 amino acids of ZNF521. Amino Acid Squence: KKRPEDGEELEDEAVHS | |
| Affinity Purified | |
| RUO | |
| Primary | |
| ZNF521 antibody is human specific. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 ug/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5 ug/mL | |
| EHZF, Evi3, LIP3, ZFP521, zinc finger protein 263, Zinc finger protein FPM315, Zinc finger protein with KRAB and SCAN domains 12 | |
| Rabbit | |
| 145 kDa | |
| 0.1 mg | |
| Immunology, Neuroscience | |
| 25925 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction