missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF521 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00 - £398.00
Specifications
| Antigen | ZNF521 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636817
|
Novus Biologicals
NBP2-41174-0.025mg |
0.025 mg |
£134.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18158578
|
Novus Biologicals
NBP2-41174 |
0.1 mg |
£398.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF521 Polyclonal specifically detects ZNF521 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| ZNF521 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| EHZF, Evi3, LIP3, ZFP521, zinc finger protein 263, Zinc finger protein FPM315, Zinc finger protein with KRAB and SCAN domains 12 | |
| ZNF521 | |
| IgG | |
| Affinity Purified | |
| 145 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Immunology, Neuroscience | |
| PBS with 0.02% Sodium Azide | |
| 25925 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human ZNF521. The immunogen is located within the first 50 amino acids of ZNF521. Amino Acid Squence: KKRPEDGEELEDEAVHS | |
| Primary | |
| ZNF521 antibody is human specific. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title