missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF502 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80130
This item is not returnable.
View return policy
Description
ZNF502 Polyclonal specifically detects ZNF502 in Human samples. It is validated for Western Blot.
Specifications
| ZNF502 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| KOX17Zinc finger protein 191, lacks zinc finger domain, Retinoic acid suppression protein A, RSG-A, Zfp191, Zinc finger and SCAN domain-containing protein 3, zinc finger protein 24, zinc finger protein 24 (KOX 17), ZNF191, ZSCAN3Zinc finger protein KOX17 | |
| Rabbit | |
| 63 kDa | |
| 100 μL | |
| Primary | |
| Equine: 85%; Rat: 83%; Canine: 77%. | |
| Human, Rat, Pig, Bovine, Canine, Equine | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_149987 | |
| ZNF502 | |
| Synthetic peptide directed towards the N terminal of human ZNF502. Peptide sequence IDSSGIVVKRFQEDEYQDSTFEEKYACEGMKENSPREIAESCLFQEGGFG. | |
| Affinity purified | |
| RUO | |
| 91392 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction