missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF428 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | ZNF428 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF428 Polyclonal specifically detects ZNF428 in Human samples. It is validated for Western Blot.Specifications
| ZNF428 | |
| Polyclonal | |
| Rabbit | |
| NP_872304 | |
| 126299 | |
| Synthetic peptide directed towards the middle region of human ZNF428. Peptide sequence PCRLCCPATAPQEAPAPEGRALGEEEEEPPRAGEGRPAGREEEEEEEEEG. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Suppressor of hairy wing homolog 1, suppressor of hairy wing homolog 1 (Drosophila), zinc finger protein 280, zinc finger protein 280A, Zinc finger protein 636,3'OY11.1SUHW1, ZNF636ZNF280 | |
| ZNF428 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title