missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF428 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80435
This item is not returnable.
View return policy
Description
ZNF428 Polyclonal specifically detects ZNF428 in Human samples. It is validated for Western Blot.
Specifications
| ZNF428 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Suppressor of hairy wing homolog 1, suppressor of hairy wing homolog 1 (Drosophila), zinc finger protein 280, zinc finger protein 280A, Zinc finger protein 636,3'OY11.1SUHW1, ZNF636ZNF280 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 126299 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_872304 | |
| ZNF428 | |
| Synthetic peptide directed towards the middle region of human ZNF428. Peptide sequence PCRLCCPATAPQEAPAPEGRALGEEEEEPPRAGEGRPAGREEEEEEEEEG. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 85%; Canine: 85%; Guinea pig: 85%. | |
| Human, Rat, Pig, Bovine, Canine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction