missing translation for 'onlineSavingsMsg'
Learn More

ZNF385A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen ZNF385A
Concentration 0.37 mg/mL
Applications Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16824805
View Documents
Proteintech
26288-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16814805
View Documents
Proteintech
26288-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF385 is a new candidate transcription factor.
Specifications

Specifications

ZNF385A
Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse
Q8VD12, Q96PM9
25946, 29813
ZNF385A Fusion Protein Ag23577
Primary
-20°C
ZNF385A
0.37 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
HZF, Retinal zinc finger protein, RZF, ZFP385, zinc finger protein 385A, ZNF385, ZNF385A
Znf385a
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.