missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF385A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26288-1-AP-20UL
This item is not returnable.
View return policy
Description
ZNF385 is a new candidate transcription factor.Specifications
| ZNF385A | |
| Polyclonal | |
| Unconjugated | |
| ZNF385A | |
| HZF, Retinal zinc finger protein, RZF, ZFP385, zinc finger protein 385A, ZNF385, ZNF385A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25946, 29813 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VD12, Q96PM9 | |
| Znf385a | |
| ZNF385A Fusion Protein Ag23577 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction