missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF273 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | ZNF273 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665551
|
Novus Biologicals
NBP2-93912-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621071
|
Novus Biologicals
NBP2-93912-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF273 Polyclonal antibody specifically detects ZNF273 in Human samples. It is validated for Western BlotSpecifications
| ZNF273 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| HZF9, ZNF273 zinc finger protein 273 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 210-260 of human ZNF273 (NP_066971.2). ECGKSCCILSQLTQHKKTATRVNFYKCKTCGKAFNQFSNLTKHKIIHPEVN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:100 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 10793 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title