missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF271 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23799-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognize both isoforms.
Zinc finger proteins interact with nucleic acids and have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins.Specifications
| ZNF271 | |
| Polyclonal | |
| Unconjugated | |
| ZNF271P | |
| ZNF271P | |
| ZNF271 Fusion Protein Ag20727 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HZF7, zinc finger protein 271, ZNF271, ZNFEB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10778 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction