missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF271 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ZNF271 |
|---|---|
| Concentration | 0.35 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16842455
|
Proteintech
23799-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16832455
|
Proteintech
23799-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody recognize both isoforms.
Zinc finger proteins interact with nucleic acids and have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins.Specifications
| ZNF271 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| HZF7, zinc finger protein 271, ZNF271, ZNFEB | |
| ZNF271P | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
| 0.35 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 10778 | |
| ZNF271 Fusion Protein Ag20727 | |
| Primary | |
| -20°C | |
| ZNF271P |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title