missing translation for 'onlineSavingsMsg'
Learn More

ZNF141 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£149.00 - £359.00

Specifications

Antigen ZNF141
Dilution Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18622889
View Documents
Novus Biologicals
NBP3-05597-100ul
100 μg
£359.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18642598
View Documents
Novus Biologicals
NBP3-05597-20ul
20 μg
£149.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF141 Polyclonal antibody specifically detects ZNF141 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

ZNF141
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
D4S90, pHZ-44, zinc finger protein 141
Recombinant fusion protein containing a sequence corresponding to amino acids 405-474 of human ZNF141 (NP_003432.1). RRSTDRSQHKKIHSADKPYKCKECDKAFKQFSLLSQHKKIHTVDKPYKCKDCDKAFKRFSHLNKHKKIHT
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
7700
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.