Learn More
RETSCH Zirconium Oxide Grinding Sets
Product Code. 10740093 Shop All Retsch Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10740093 100 mL Each
10094911 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10740093 Supplier RETSCH Supplier No. 01.462.0192
Only null left
Add to basket
Add to basket
Designed for extremely quick, loss-free and reproducible grinding of medium-hard, brittle and fibrous materials to analytical fineness. Retsch™ Zirconium Oxide Grinding Sets are ideal for use in the building materials sector (cement), in geology, mineralogy, metallurgy and in power plants.
  • For use with RS 100 and RS 200 Vibratory Disc Mills
Specifications
Product Type Grinding Set
Capacity (Metric) 100 mL
For Use With (Equipment) For RS200
Material Zirconium Oxide