Learn More
RETSCH Zirconium Oxide Grinding Sets
Designed for extremely quick, loss-free and reproducible grinding of medium-hard, brittle and fibrous materials to analytical fineness. Retsch™ Zirconium Oxide Grinding Sets are ideal for use in the building materials sector (cement), in geology, mineralogy, metallurgy and in power plants.
Brand:  RETSCH 01.462.0193
Code : A0
Additional Details : Weight : 6.00000kg
 View more versions of this product
Product Code. 10094911
  • £5530.00 / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
  • For use with RS 100 and RS 200 Vibratory Disc Mills
Specifications
Specifications
Grinding Set
50 mL
For RS200
Zirconium Oxide