missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Zinc phthalocyanine

Product Code. 16461779
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461779 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16461779 Supplier MedChemExpress Supplier No. HY192041g
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Zinc phthalocyanine is commonly applied in industry (catalysts, photoconductors) and biomedical (photodynamic therapy, PDT). Zinc phthalocyanine can be used to photooxidise cyclohexane.

Specifications

Content And Storage 4°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen)
Product Type Dye Reagents
Quantity 1 g
Shipping Condition Room temperature
Solubility DMF : 2.7 mg/mL (4.67 mM; Need ultrasonic and warming)
Target HBV
Molecular Weight (g/mol) 577.9 g/mol
CAS 14320-04-8
Synonym ZnPc
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.