Learn More
Zinc oxalate dihydrate, Puratronic™, 99.999% (metals basis)
Description
Zinc oxalate dihydrate is used as an chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Molecular Formula | C2H4O6Zn |
| Linear Formula | ZnC2O4·2H2O |
| UN Number | UN3288 |
| Merck Index | 14,10146 |
| Synonym | 5-chloro-2-methoxyaniline hydrochloride, hindamine red r, azobase cas, hindasol red rc, stabamine red rc, devol red j, fast red r base, red base ciba i, red base irga i, red salt ciba i |
| Solubility Information | Soluble in dilute mineral acids. |
| InChI Key | WCKIDCVWRJUPFY-UHFFFAOYSA-L |
| Show More |
Safety and Handling
- Zinc oxalate dihydrate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contain: Zinc oxalate dihydrate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.