accessibility menu, dialog, popup

UserName

OOOOH2OH2OZn

CAS RN 55906-21-3

IUPAC Name: zinc(2+) dihydrate oxalate
Synonyms: zinc(2+) dihydrate oxalate
Molecular Weight (g/mol): 189.43
Molecular Formula: C2H4O6Zn
InChi Key: WCKIDCVWRJUPFY-UHFFFAOYSA-L
SMILES: O.O.[Zn++].[O-]C(=O)C([O-])=O
Grade 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results