missing translation for 'onlineSavingsMsg'
Learn More

Zinc hexafluoro-2,4-pentanedionate dihydrate

Product Code. 11338378
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338378 1 g 1g
11328378 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338378 Supplier Thermo Scientific Alfa Aesar Supplier No. 039559.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16743-33-2 | C10H6F12O6Zn | 515.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16743-33-2
Molecular Formula C10H6F12O6Zn
Molecular Weight (g/mol) 515.51
MDL Number MFCD00150577
InChI Key HHDNHEKLDWSJAJ-LJDKTGGESA-L
Synonym zinc hexafluoroacetylacetonate hydrate, zinc hexafluoroacetylacetonate dihydrate, zinc hexafluoro 2,4-pentanedionate, zinc hexafluoro-2,4-pentanedionate dihydrate, hexafluoroacetylacetone zinc derivative dihydrate, 3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy zincio oxy pent-3-en-2-one dihydrate
PubChem CID 72376390
SMILES O.O.[Zn++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 157°C to 158°C
Physical Form Powder
Quantity 1 g
IUPAC Name zinc(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate) dihydrate
Formula Weight 515.49 (479.47 Anhydrous)
Chemical Name or Material Zinc hexafluoro-2,4-pentanedionate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.