accessibility menu, dialog, popup

UserName

OOOOZnH2OH2OCF3CF3CF3CF3

CAS RN 16743-33-2

IUPAC Name: bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) dihydrate zinc
Synonyms: bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) dihydrate zinc
Molecular Weight (g/mol): 517.53
Molecular Formula: C10H8F12O6Zn
InChi Key: KPMCYOMCJRBFLE-UHFFFAOYSA-N
SMILES: O.O.[Zn].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results