Learn More
Zinc cyanide, 98%, pure, Thermo Scientific Chemicals
£13.60 - £79.00
Chemical Identifiers
CAS | 557-21-1 |
---|---|
Molecular Formula | C2N2Zn |
Molecular Weight (g/mol) | 117.43 |
MDL Number | MFCD00011292 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10203012
|
Thermo Scientific Acros
368070050 |
5 g | Glass Bottle |
£13.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10532072
|
Thermo Scientific Acros
368072500 |
250 g | Plastic bottle |
£28.30
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10592242
|
Thermo Scientific Acros
368070010 |
1 kg | Plastic bottle |
£79.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
557-21-1 | |
C2N2Zn | |
117.43 | |
MFCD00011292 |
Specifications
800.0°C | |
White | |
97.5% min. (Complexometry) | |
Zn(CN)2 | |
02,III,86 | |
Solubility in water: insoluble | |
117.43 | |
Pure |
557-21-1 | |
Powder | |
C2N2Zn | |
MFCD00011292 | |
15,10334 | |
117.43 | |
98% | |
Zinc cyanide, 98% |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
Contact with acids liberates very toxic gas.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Keep container tightly closed.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-162-9
- Zinc cyanide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H310-Fatal in contact with skin.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H330-Fatal if inhaled.
- H300-Fatal if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P233-Keep container tightly closed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P361-Take off immediately all contaminated clothing.
RUO – Research Use Only