missing translation for 'onlineSavingsMsg'
Learn More

Zinc cyanide, 98%, pure

Product Code. 10592242 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
250g
5g
This item is not returnable. View return policy

Product Code. 10592242

Brand: Thermo Scientific Acros 368070010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 557-21-1 | C2N2Zn | 117.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-21-1
Molecular Formula C2N2Zn
Molecular Weight (g/mol) 117.43
MDL Number MFCD00011292

Specifications

Melting Point 800.0°C
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula Zn(CN)2
Beilstein 02,III,86
Merck Index 15,10334
Solubility Information Solubility in water: insoluble
Formula Weight 117.43
Percent Purity 98%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Zinc cyanide, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Zinc cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.