missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus (SPH2015) NS1 Mouse anti-Virus, HRP, Antibody Pair, Novus Biologicals™
Antibody pair for Solid phase sandwich ELISA
£342.00 - £832.00
Specifications
| Antigen | Zika Virus (SPH2015) NS1 |
|---|---|
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18389721
|
Novus Biologicals
NBP3-06710-5PLATES |
5 Plates |
£342.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18357413
|
Novus Biologicals
NBP3-06710-15PLATES |
15 Plates |
£832.00
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Solid phase sandwich ELISA for quantitative determination of Zika Virus NS1.Specifications
| Zika Virus (SPH2015) NS1 | |
| Monoclonal | |
| Mouse | |
| Virus | |
| Storage of components varies. See protocol for specific instructions. |
| ELISA | |
| HRP | |
| RUO | |
| Zika NS1, ZIKV NS1, ZIKV-NS1 | |
| Antibody Pairs |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title