missing translation for 'onlineSavingsMsg'
Learn More

Zika Virus (SPH2015) NS1 Mouse anti-Virus, HRP, Antibody Pair, Novus Biologicals™

Product Code. 18357413 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357413 15 Plates Pack of 15
18389721 5 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18357413 Supplier Novus Biologicals Supplier No. NBP30671015PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody pair for Solid phase sandwich ELISA

Solid phase sandwich ELISA for quantitative determination of Zika Virus NS1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus (SPH2015) NS1
Applications ELISA
Classification Monoclonal
Conjugate HRP
Gene Alias Zika NS1, ZIKV NS1, ZIKV-NS1
Host Species Mouse
Quantity 15 Plates
Regulatory Status RUO
Target Species Virus
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Type Antibody Pairs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.