missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus (SPH2015) Envelope anti-Virus, HRP, Antibody Pair, Novus Biologicals™
Antibody pair for Solid phase sandwich ELISA
£348.00 - £832.00
Specifications
| Antigen | Zika Virus (SPH2015) Envelope |
|---|---|
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18389571
|
Novus Biologicals
NBP3-06709-5PLATES |
5 Plates |
£348.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18388442
|
Novus Biologicals
NBP3-06709-15PLATES |
15 Plates |
£832.00
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Solid phase sandwich ELISA for quantitative determination of Zika Virus Envelope.Specifications
| Zika Virus (SPH2015) Envelope | |
| Monoclonal | |
| RUO | |
| Zika Envelope | |
| Antibody Pairs |
| ELISA | |
| HRP | |
| Virus | |
| Storage of components varies. See protocol for specific instructions. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel