missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus (SPH2015) Envelope anti-Virus, HRP, Antibody Pair, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Solid phase sandwich ELISA for quantitative determination of Zika Virus Envelope.
Specifications
Specifications
| Antigen | Zika Virus (SPH2015) Envelope |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Gene Alias | Zika Envelope |
| Quantity | 15 Plates |
| Regulatory Status | RUO |
| Target Species | Virus |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Product Type | Antibody Pairs |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?