missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE9 Antibody (SR1275), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | ZFYVE9 |
|---|---|
| Clone | SR1275 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679134
|
Novus Biologicals
NBP3-22552-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632575
|
Novus Biologicals
NBP3-22552-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZFYVE9 Monoclonal antibody specifically detects ZFYVE9 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| ZFYVE9 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Madh-interacting protein, MADHIP, mothers against decapentaplegic homolog interacting protein, mothers against decapentaplegic homolog interacting protein, receptoractivation anchor, NSP, receptor activation anchor, SARAhSARA, Smad anchor for receptor activation, SMADIPMAD, mothers against decapentaplegic homolog (Drosophila) interacting protein, zinc finger FYVE domain-containing protein 9, zinc finger, FYVE domain containing 9 | |
| A synthesized peptide derived from human ZFYVE9 (Uniprot #: O95405) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1275 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 9372 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title