missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE9 Antibody (SR1275), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22552-100ul
This item is not returnable.
View return policy
Description
ZFYVE9 Monoclonal antibody specifically detects ZFYVE9 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| ZFYVE9 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1275 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Madh-interacting protein, MADHIP, mothers against decapentaplegic homolog interacting protein, mothers against decapentaplegic homolog interacting protein, receptoractivation anchor, NSP, receptor activation anchor, SARAhSARA, Smad anchor for receptor activation, SMADIPMAD, mothers against decapentaplegic homolog (Drosophila) interacting protein, zinc finger FYVE domain-containing protein 9, zinc finger, FYVE domain containing 9 | |
| A synthesized peptide derived from human ZFYVE9 (Uniprot #: O95405) | |
| 100 μg | |
| Signal Transduction | |
| 9372 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction