missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP36 Monoclonal Antibody (K94030_2F8), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | ZFP36 |
|---|---|
| Clone | K94030_2F8 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30203548
|
Invitrogen™
MA555705 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30205203
|
Invitrogen™
MA556085 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: DLAAPGHPPV LRQSISFSGL PSGRRTSPPP PGLAGPSLSS SSFSPSSSPP PPGDLPLSPS AFSAAPGTPL ARRDPTPVCC PSCRRATPIS VWGPLGGLVR TPSVQSLGSD PDEYASSGSS LGGSDSPVFE AGVFAPPQPV
ZFP36 Monoclonal antibody specifically detects ZFP36 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| ZFP36 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| G0/G1 switch regulatory protein 24; G0S24; GOS24; growth factor-inducible nuclear protein NUP475; mRNA decay activator protein ZFP36; Nup475; RNF162A; Tax interacting protein; TIS11; TIS11A; TIS11D; TISII; TNF alpha inhibitor; TPA-induced sequence 11; Tristetraprolin; tristetraprolin; LOW QUALITY PROTEIN: mRNA decay activator protein ZFP36; tristetraproline; TTP; Zfp36; zfp-36; ZFP36 ring finger protein; zinc finger protein 36; zinc finger protein 36 homolog; zinc finger protein 36, C3H type, homolog; zinc finger protein, C3H type, 36 homolog | |
| Recombinant human ZFP36 (immunogen range is 171-310). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Zfp36 |
| K94030_2F8 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P26651 | |
| 7538 | |
| IgG2a | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title