missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZFP36 Monoclonal Antibody (K94030_2F8), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556085

 View more versions of this product

Product Code. 30205203

  • £734.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: DLAAPGHPPV LRQSISFSGL PSGRRTSPPP PGLAGPSLSS SSFSPSSSPP PPGDLPLSPS AFSAAPGTPL ARRDPTPVCC PSCRRATPIS VWGPLGGLVR TPSVQSLGSD PDEYASSGSS LGGSDSPVFE AGVFAPPQPV

ZFP36 is a probable regulatory protein with a novel zinc finger structure involved in regulating the response to growth factors. Knockdown of ZFP36 increased both cognate macrophage gene mRNAs and inflammatory tumor necrosis factor protein release. Overexpression of ZFP36 resulted in decreased levels of the same genes supporting its role in regulating macrophage gene expression. Multiple phosphorylation sites in ZFP36 in mammalian cells were reported. ZFP36 can be phosphorylated by JNK as well as by the other members of the greater MAP kinase family. Gene expression profiling predicts clinical outcome of prostate cancer. Inappropriate ZFP36 production may be one factor that contributes to higher rheumatoid arthritis disease activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZFP36
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
P26651
Zfp36
Recombinant human ZFP36 (immunogen range is 171-310).
200 μL
Primary
Human
Antibody
IgG2a
Immunoprecipitation, Western Blot
K94030_2F8
Unconjugated
Zfp36
G0/G1 switch regulatory protein 24; G0S24; GOS24; growth factor-inducible nuclear protein NUP475; mRNA decay activator protein ZFP36; Nup475; RNF162A; Tax interacting protein; TIS11; TIS11A; TIS11D; TISII; TNF alpha inhibitor; TPA-induced sequence 11; Tristetraprolin; tristetraprolin; LOW QUALITY PROTEIN: mRNA decay activator protein ZFP36; tristetraproline; TTP; Zfp36; zfp-36; ZFP36 ring finger protein; zinc finger protein 36; zinc finger protein 36 homolog; zinc finger protein 36, C3H type, homolog; zinc finger protein, C3H type, 36 homolog
Mouse
Protein A
RUO
7538
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.