missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zenon™ pHrodo™ iFL IgG Labeling Reagents

Product Code. 15871527
Change view
Click to view available options
Quantity:
250 μL
1.2 mL
Species:
Human
Mouse
Label or Dye:
pHrodo Deep Red
pHrodo Green
pHrodo Red
pHrodo iFL Green
pHrodo iFL Red
Color:
Deep Red
Green
Red
Unit Size:
1.2mL
240µL
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Species Label or Dye Color unitSize
15871527 250 μL Human pHrodo iFL Green Green Each
17964691 250 μL Human pHrodo Deep Red Deep Red 240µL
17924701 250 μL Mouse pHrodo Deep Red Deep Red 240µL
16099321 250 μL Human pHrodo iFL Red Red 240µL
16344925 250 μL Mouse pHrodo iFL Red Red 240µL
16334925 250 μL Mouse pHrodo iFL Green Green 240µL
30534651 1.2 mL Human pHrodo Deep Red Deep Red 1.2mL
17865247 1.2 mL Human pHrodo iFL Red Red 1.2mL
17835097 1.2 mL Human pHrodo iFL Green Green 1.2mL
30535648 1.2 mL Mouse pHrodo Deep Red Deep Red 1.2mL
30535017 1.2 mL Mouse pHrodo Red Red 1.2mL
30535977 1.2 mL Mouse pHrodo Green Green 1.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 15871527 Supplier Invitrogen™ Supplier No. Z25611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex.

Streamline antibody-drug conjugate screening campaigns with Invitrogen Zenon pHrodo IgG Labeling Reagents to screen for antibody internalization. These reagents provide rapid, scalable antibody labeling and give a bright fluorescence-based indication of antibody uptake and trafficking in target cells when analyzed by flow cytometry, fluorescence microscopy, or high-content screening.

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex. Formation of the Fab–antibody complex occurs in fewer than 5 minutes. Because the pHrodo™ iFL dyes dramatically increase in fluorescence as the pH of their surroundings becomes more acidic, the antibodies coupled with the Zenon™ pHrodo™ iFL IgG Labeling Reagents provide an excellent measure of endocytic activity based on the acidification of the labeled antibodies as they are ingested by the cell.

The Zenon™ pHrodo™ iFL IgG Labeling Reagents contain sufficient material to label one to four 96-well plates, depending on the Zenon™ labeling reagent concentration used. Only a small amount of starting material is needed, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions.

Other advantages of the pH-sensitive Zenon™ pHrodo™ iFL IgG Labeling Reagents include:

  • Simplicity—No reagent purification is required
  • Accuracy—No wash steps or quenchers are needed, minimizing cross-reactivity or background
  • Speed—Labeled antibodies are typically ready to use in 10 minutes
  • Specificity—Dyes are designed for the specific detection of phagocytosis and endocytosis

Preserve primary antibody function and affinities

Reactive dye labeling of primary antibodies can have unpredictable and undesirable outcomes. One of these is reduced binding affinity due to label addition in the binding pocket. The Zenon™ antibody labeling approach, which is targeted to the Fc tail, resolves this potential issue.

Try custom antibody conjugates

If the antibody conjugate you need is not available as one of our stocked products, we will prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Order Info

Store in refrigerator at 2°C to 8°C.

TRUSTED_SUSTAINABILITY

Specifications

Description pHrodo™ iFL Green, 240 μL
Kit Contents 1 vial of Zenon™ pHrodo™ iFL Green human labeling reagent (250 μL)
Product Type Labeling Reagent
Quantity 250 μL
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light
Excitation/Emission 505/530 nm
Product Line Zenon
Label or Dye pHrodo iFL Green
Color Green
Labeling Target IgG
Species Human
Label Type pHrodo™ (pH sensitive)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.