missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Zenon™ Alexa Fluor™ 405 Mouse IgG2a Labeling Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
Zenon labeling technology provides a fast, versatile and reliable method for producing antibody conjugates, even with very small (submicrogram) amounts of starting material. Antibody conjugates formed using Zenon technology may be used to stain cells in any protocol where a directly labeled primary antibody is suitable, including flow cytometry, imaging, high throughput and other applications. Moreover, this technology simplifies applications that previously were time consuming or not practical, such as the use of multiple mouse-derived antibodies in the same staining protocol.
View a selection guide for all Zenon™ antibody labeling kits and other antibody labeling products.
View a selection guide for all Zenon™ antibody labeling kits and other antibody labeling products.
Specifications
Specifications
| Description | Zenon™ Alexa Fluor™ 405 Mouse IgG2a Labeling Kit, 1 Kit, 421nm Emission, 401nm Excitation, 2 to 8°C Storage Requirements, Used for Labeling Chemistry, Protein Labeling and Crosslinking |
| For Use With (Application) | Labeling chemistry, protein labeling and crosslinking, protein biology, cell analysis, protein, peptide and antibody labeling |
| Quantity | 1 Kit |
| Product Type | Zenon™ Alexa Fluor™ 405 Mouse IgG2a Labeling Kit |
| Kit Contents | 1 Vial of Zenon™ Alexa Fluor™ 405 mouse IgG2a labeling reagent (250μL) and 1 Vial of Zenon™ blocking reagent (mouse IgG, 250μL) |
| Content And Storage | 2°C to 8°C |
| Label or Dye | Alexa Fluor™ 405 |
| Emission | 421 |
| Excitation | 401 |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction